Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

759236

Sigma-Aldrich

1-(2-Bromophenyl)piperidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14BrN
CAS Number:
Molecular Weight:
240.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.580

density

1.339 g/mL at 25 °C

functional group

bromo

SMILES string

Brc1ccccc1N2CCCCC2

InChI

1S/C11H14BrN/c12-10-6-2-3-7-11(10)13-8-4-1-5-9-13/h2-3,6-7H,1,4-5,8-9H2

Inchi Key

ULDSFDQNVQGLCC-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service