Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

758442

Sigma-Aldrich

1,2,3,4,5,6,7,8-Octafluoro-9,10-bis[2-(2,4,6-trimethylphenyl)ethynyl]anthracene

97%

Synonym(s):

1,2,3,4,5,6,7,8-Octafluoro-9,10-bis(mesitylethynyl)anthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H22F8
CAS Number:
Molecular Weight:
606.55
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

powder

mp

267-272 °C

orbital energy

HOMO 5.7 eV (CV)
LUMO 3.5 eV (CV)

SMILES string

Cc1cc(C)c(C#Cc2c3c(F)c(F)c(F)c(F)c3c(C#Cc4c(C)cc(C)cc4C)c5c(F)c(F)c(F)c(F)c25)c(C)c1

InChI

1S/C36H22F8/c1-15-11-17(3)21(18(4)12-15)7-9-23-25-27(31(39)35(43)33(41)29(25)37)24(10-8-22-19(5)13-16(2)14-20(22)6)28-26(23)30(38)34(42)36(44)32(28)40/h11-14H,1-6H3

InChI key

YYIVDNHHWCQYCH-UHFFFAOYSA-N

Application

This material is a relatively new candidate as a n-type semiconductor for use in Organic Field Effect Transistor (OFET) devices.[1] It displays very impressive fluorescence useful in the field of optics research. The ethynyl bridges allow the molecule to stack more efficiently allowing for enhanced electron transport.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John F Tannaci et al.
The Journal of organic chemistry, 73(20), 7895-7900 (2008-09-16)
9,10-Dichlorooctafluoroanthracene (1) reacts with aryl boronic acids and terminal alkynes under palladium-catalyzed cross-coupling conditions to afford 9,10-diaryloctafluoroanthracenes (2a-e) and 9,10-dialkynyloctafluoroanthracenes (6a,b), respectively. Optical spectroscopy and cyclic voltammetry indicate that octafluoro-9,10-di(thiophen-2-yl)anthracene (2d) exhibits donor-acceptor character and a LUMO energy level of

Articles

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service