Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757284

Sigma-Aldrich

4,4′-Cyclohexylidenebis[N,N-bis(4-methylphenyl)benzenamine]

97%

Synonym(s):

TAPC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H46N2
CAS Number:
Molecular Weight:
626.87
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

powder

mp

180-185 °C

orbital energy

HOMO 5.5 eV 
LUMO 2 eV 

OLED device performance

ITO/TAPC/Alq3/Mg:Ag

  • Color: green
  • Max. Luminance: >1,000 Cd/m2
  • Turn-On Voltage: 2.5 V

ITO/TAPC/CBP:FIrpic (6%)/PO14/LiF/Al
  • Color: blue
  • Max. EQE: 12.5 %

ITO/TAPC/CBP:Ir(ppy)2(acac) (6%)/BCP/LiF/Al
  • Color: green
  • Max. EQE: 13 %

ITO/TAPC/CBP:Ir(ppy)2(acac) (6%)/PO14/LiF/Al
  • Color: green
  • Max. EQE: 16 %

SMILES string

Cc1ccc(cc1)N(c2ccc(C)cc2)c3ccc(cc3)C4(CCCCC4)c5ccc(cc5)N(c6ccc(C)cc6)c7ccc(C)cc7

InChI

1S/C46H46N2/c1-34-8-20-40(21-9-34)47(41-22-10-35(2)11-23-41)44-28-16-38(17-29-44)46(32-6-5-7-33-46)39-18-30-45(31-19-39)48(42-24-12-36(3)13-25-42)43-26-14-37(4)15-27-43/h8-31H,5-7,32-33H2,1-4H3

InChI key

ZOKIJILZFXPFTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This material is a popular Hole Transport Layer material for OLED devices.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Libby O Onyeka et al.
Journal of food protection, 83(3), 476-484 (2020-02-18)
A cross-sectional study was conducted to determine the prevalence of and factors associated with Shiga toxin-producing Escherichia coli (STEC) in raw beef and ready-to-eat (RTE) beef products sold in 31 retail outlets in Pretoria, South Africa, and nearby areas. A
Mei Yi Sim et al.
Life sciences, 203, 282-290 (2018-04-25)
To investigate the effect of dioxonaphthoimidazolium analog YM155 on cell cycle progression of the clear-cell variant of renal cell carcinoma (ccRCC). Cell cycle analysis was performed using bromodeoxyuridine (BrdU) and PI, apoptosis initiation was monitored using Annexin V and proteins
Maria Lúcia Guerra Monteiro et al.
Scientific reports, 8(1), 9524-9524 (2018-06-24)
This study investigated the effects of high hydrostatic pressure (HHP) and ultraviolet radiation (UV-C), individually and combined, on the physical, chemical and bacterial parameters of Nile tilapia (Oreochromis niloticus) fillets stored at 4 °C for 14 days. Tilapia fillets were divided
Organic electroluminescent diodes
Tang, C. W.; et al.
Applied Physics Letters, 51, 913-913 (1987)
César A Lázaro et al.
Molecules (Basel, Switzerland), 25(14) (2020-07-19)
This study investigated the isolated effect of modified atmosphere packaging (MAP; 50% CO2 and 50% N2) and ultraviolet radiation (UV; 0.30 J/cm2) as well as their combined (MAP/UV) effect on reduction of Salmonella typhimurium and Escherichia coli O157:H7, biogenic amines

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service