Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755818

Sigma-Aldrich

2-Hydroxy-3-{3-[2,4,6,8-tetramethyl-4,6,8-tris(propyl glycidyl ether)-2-cyclotetrasiloxanyl]propoxy}propyl methacrylate

Synonym(s):

CS-783, Methacrylate epoxy cyclosiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H62O14Si4
CAS Number:
Molecular Weight:
783.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

refractive index

n20/D 1.468

viscosity

220 cP(25 °C)

density

1.130 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCC(O)COCCC[Si]1(C)O[Si](C)(CCCOCC2CO2)O[Si](C)(CCCOCC3CO3)O[Si](C)(CCCOCC4CO4)O1

InChI

1S/C32H62O14Si4/c1-27(2)32(34)42-20-28(33)19-35-11-7-15-47(3)43-48(4,16-8-12-36-21-29-24-39-29)45-50(6,18-10-14-38-23-31-26-41-31)46-49(5,44-47)17-9-13-37-22-30-25-40-30/h28-31,33H,1,7-26H2,2-6H3

InChI key

MDIPNFHXQNLQFC-UHFFFAOYSA-N

General description

  • ·Dual cure mechanism
  • ·Multifunctional
  • ·UV curable
  • ·Low chloride
  • ·Low viscosity

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>399.9 °F

flash_point_c

> 204.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service