Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755001

Sigma-Aldrich

(S)-2-(Trifluoromethyl)piperidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10F3N
CAS Number:
Molecular Weight:
153.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

optical activity

[α]22/D -14.4°, c = 0.5% in methanol

refractive index

n20/D 1.391

density

1.161 g/mL at 25 °C

functional group

fluoro

SMILES string

FC(F)(F)[C@@H]1CCCCN1

InChI

1S/C6H10F3N/c7-6(8,9)5-3-1-2-4-10-5/h5,10H,1-4H2/t5-/m0/s1

InChI key

NAXDEFXCCITWEU-YFKPBYRVSA-N

Application

(S)-2-(Trifluoromethyl)piperidine can be used as a reactant for the synthesis of (1R,2R)-2-((S)-2-(trifluoromethyl)piperidin-1-yl)-1,2-dihydronaphthalen-1-ol by ring-opening reaction of oxabenzonorbornadiene in the presence of iridium catalyst.[1]

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

79.0 °F - closed cup

flash_point_c

26.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water-Promoted Ir-Catalyzed Ring-Opening of Oxa (aza) benzonorbornadienes with Fluoroalkylamines
Yang X, et al.
The Journal of Organic Chemistry, 83(17), 10097-10106 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service