Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

752916

Sigma-Aldrich

Octafluoroanthraquinone

96%

Synonym(s):

1,2,3,4,5,6,7,8-Octafluoro-9,10-anthracenedione, 1,2,3,4,5,6,7,8-Octafluoro-9,10-anthraquinone, Octafluoro-9,10-anthraquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14F8O2
CAS Number:
Molecular Weight:
352.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

96%

form

powder or crystals

mp

270-300 °C

SMILES string

Fc1c(F)c(F)c2C(=O)c3c(F)c(F)c(F)c(F)c3C(=O)c2c1F

InChI

1S/C14F8O2/c15-5-1-2(6(16)10(20)9(5)19)14(24)4-3(13(1)23)7(17)11(21)12(22)8(4)18

InChI key

JHJWVVOLGHMBPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John F Tannaci et al.
The Journal of organic chemistry, 72(15), 5567-5573 (2007-06-26)
9,10-Dichlorooctafluoroanthracene (1) was synthesized from commercially available tetrafluorophthalic acid by an optimized solution-phase route. To establish 1 as a synthon for n-type organic semiconductors, the compound was reacted with phenylboronic acid under modified Suzuki-Miyaura coupling conditions to generate octafluoro-9,10-diphenylanthracene (7)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service