Skip to Content
MilliporeSigma
All Photos(3)

Documents

750662

Sigma-Aldrich

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II)

97%

Synonym(s):

Lead bis(2,2,6,6-tetramethyl-3,5-heptanedionate), Pb(TMHD)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(OCC(CH3)3CHCOC(CH3)3)2
CAS Number:
Molecular Weight:
573.73
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

core: lead
reagent type: catalyst

mp

129-131 °C (lit.)
129-133 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)C(=O)\C=C(/O[PbH2]O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/2C11H20O2.Pb/c2*1-10(2,3)8(12)7-9(13)11(4,5)6;/h2*7,12H,1-6H3;/q;;+2/p-2/b2*8-7-;

InChI key

RFDHUANRWFSKRW-ATMONBRVSA-L

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

CVD precursor for the manufacture of advanced dielectric and ferroelectric thing films. Used for lead oxide and mixed-metal thin films growth by a variety of deposition methods. Shows superior chemical compatibility and improved safety in MOCVD of PNZT.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service