Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

748765

Sigma-Aldrich

Isopropyl 1H-imidazole-1-carboxylate

95%

Synonym(s):

Heller-Sarpong reagent, IPImC, Imidazole-1-carboxylic acid isopropyl ester, Isopropyl imidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O2
CAS Number:
Molecular Weight:
154.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.467

density

1.098 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)OC(=O)n1ccnc1

InChI

1S/C7H10N2O2/c1-6(2)11-7(10)9-4-3-8-5-9/h3-6H,1-2H3

InChI key

UAILNBAKJABHNQ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

226.4 °F

flash_point_c

108 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Professor Heller's research focuses on mild acylation reactions using carbonylimidazole-derived reagents.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service