Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

744999

Sigma-Aldrich

Dichlorobis[tri(1-piperidinyl)phosphine]palladium(II)

≥97.0% (CHN)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H60Cl2N6P2Pd
CAS Number:
Molecular Weight:
744.11
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (CHN)

form

powder

composition

carbon, 47.0-49.9%

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

suitability

complies for Infrared spectrum

SMILES string

Cl[Pd]Cl.C1CCN(CC1)P(N2CCCCC2)N3CCCCC3.C4CCN(CC4)P(N5CCCCC5)N6CCCCC6

InChI

1S/2C15H30N3P.2ClH.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*1-15H2;2*1H;/q;;;;+2/p-2

Inchi Key

GNTXTSSDJRUYPK-UHFFFAOYSA-L

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mizoroki-Heck reactions catalyzed by palladium dichloro-bis(aminophosphine) complexes under mild reaction conditions. The importance of ligand composition on the catalytic activity.
Oberholzer, M. and Frech, C.M.
Green Chemistry, 15(6), 1678-1678 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service