Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

743933

Sigma-Aldrich

1-[2-Oxo-2-(2-pyridinyl)ethyl]pyridinium iodide

Synonym(s):

1-(2-Pyridacyl)pyridinium iodide, 1-(2-Pyridylcarbonylmethyl)pyridinium iodide, 1-[2-(2-Pyridinyl)-2-oxoethyl]pyridinium iodide, Kröhnke reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11IN2O
CAS Number:
Molecular Weight:
326.13
Beilstein/REAXYS Number:
3577311
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97.5-102.5% (AT)

form

solid

reaction suitability

reaction type: C-C Bond Formation

SMILES string

[I-].O=C(C[n+]1ccccc1)c2ccccn2

InChI

1S/C12H11N2O.HI/c15-12(11-6-2-3-7-13-11)10-14-8-4-1-5-9-14;/h1-9H,10H2;1H/q+1;/p-1

InChI key

BWAWQRQYQUSQEV-UHFFFAOYSA-M

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service