Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

741280

Sigma-Aldrich

3-(Trifluoromethyl)pyridazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3F3N2
CAS Number:
Molecular Weight:
148.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

30-35 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cccnn1

InChI

1S/C5H3F3N2/c6-5(7,8)4-2-1-3-9-10-4/h1-3H

InChI key

VVQHIHPZHJQYEY-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

204.8 °F

flash_point_c

96 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service