Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740616

Sigma-Aldrich

9-Azajulolidine

97%

Synonym(s):

2,3,6,7-Tetrahydro-1H,5H-9-azabenzo[ij]quinolizine, 4,5,6,8,9,10-Hexahydropyrido[3,2,1-ij][1,6]naphthyridine, 5,6,9,10-Tetrahydro-4H,8H-pyrido-[3,2,1-ij][1,6]naphthyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2
CAS Number:
Molecular Weight:
174.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

97%

mp

70-74 °C

storage temp.

2-8°C

SMILES string

C1CN2CCCc3cncc(C1)c23

InChI

1S/C11H14N2/c1-3-9-7-12-8-10-4-2-6-13(5-1)11(9)10/h7-8H,1-6H2

InChI key

VARHFUNXFXTFII-UHFFFAOYSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service