Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740128

Sigma-Aldrich

1,1-Dichloro-2-vinylcyclopropane

95%

Synonym(s):

1,1-Dichloro-2-vinylcyclopropane, 1,1-Dichloro-2-ethenylcyclopropane, 2,2-Dichlorocyclopropylethene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6Cl2
CAS Number:
Molecular Weight:
137.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.474

density

1.151 at 25 °C

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC1(Cl)CC1C=C

InChI

1S/C5H6Cl2/c1-2-4-3-5(4,6)7/h2,4H,1,3H2

InChI key

GAAAGAHNGGKSQH-UHFFFAOYSA-N

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

84.9 °F

flash_point_c

29.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service