Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

734969

Sigma-Aldrich

Methyl thiophene-2-carboxylate

97%

Synonym(s):

Methyl thiophene-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2S
CAS Number:
Molecular Weight:
142.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

liquid

density

1.219 g/mL at 25 °C

SMILES string

COC(=O)c1cccs1

InChI

1S/C6H6O2S/c1-8-6(7)5-3-2-4-9-5/h2-4H,1H3

Inchi Key

PGBFYLVIMDQYMS-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

330.8 °F

flash_point_c

166 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service