Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731307

Sigma-Aldrich

3-Bromo-1-(phenylsulfonyl)-7-azaindole

96%

Synonym(s):

3-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9BrN2O2S
Molecular Weight:
337.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

solid

mp

140-145 °C

storage temp.

2-8°C

SMILES string

Brc1cn(c2ncccc12)S(=O)(=O)c3ccccc3

InChI

1S/C13H9BrN2O2S/c14-12-9-16(13-11(12)7-4-8-15-13)19(17,18)10-5-2-1-3-6-10/h1-9H

InChI key

TZTSMNAZKFSLIC-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service