About This Item
assay
≥99.0% (GC)
refractive index
n20/D 1.536 (lit.)
bp
231-232 °C (lit.)
density
0.922 g/mL at 25 °C (lit.)
SMILES string
CCN(CC)c1cccc(C)c1
InChI
1S/C11H17N/c1-4-12(5-2)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3
InChI key
CIPVVROJHKLHJI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 2
flash_point_f
212.0 °F - closed cup
flash_point_c
100 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service