Skip to Content
MilliporeSigma
All Photos(1)

Documents

727253

Sigma-Aldrich

(R)-(3-Fluorophenyl)oxirane

ChiPros®, produced by BASF, ≥98.0%

Synonym(s):

(R)-3-Fluorostyrene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7FO
CAS Number:
Molecular Weight:
138.14
Beilstein/REAXYS Number:
9694875
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

produced by BASF

assay

≥98.0% (GC)
≥98.0%

form

liquid

optical purity

enantiomeric excess: ≥98.0%

SMILES string

Fc1cccc(c1)[C@@H]2CO2

InChI

1S/C8H7FO/c9-7-3-1-2-6(4-7)8-5-10-8/h1-4,8H,5H2/t8-/m0/s1

InChI key

HNBRZCKMGQHNJA-QMMMGPOBSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

pictograms

Health hazardSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Carc. 1B - Eye Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service