Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

723800

Sigma-Aldrich

3-Ethyl-1-pentyn-3-ol

98.0%

Synonym(s):

(1-Ethyl-1-hydroxypropyl)acetylene, 3-Ethylpent-1-yn-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH≡CC(OH)(CH2CH3)2
CAS Number:
Molecular Weight:
112.17
Beilstein/REAXYS Number:
1740944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98.0%

form

liquid

bp

138-140 °C (lit.)

density

0.872 g/mL at 20 °C (lit.)

SMILES string

CCC(O)(CC)C#C

InChI

1S/C7H12O/c1-4-7(8,5-2)6-3/h1,8H,5-6H2,2-3H3

InChI key

PUNRPAWKFTXZIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

98.6 °F - closed cup

flash_point_c

37 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service