Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

721220

Sigma-Aldrich

2-Methyl-6-(tributylstannyl)pyridine

Synonym(s):

2-Methyl-6-tributylstannanylpyridine, 6-Methylpyridin-2-yltributylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H33NSn
CAS Number:
Molecular Weight:
382.17
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.512

density

1.134 g/mL at 25 °C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1cccc(C)n1

InChI

1S/C6H6N.3C4H9.Sn/c1-6-4-2-3-5-7-6;3*1-3-4-2;/h2-4H,1H3;3*1,3-4H2,2H3;

InChI key

USKQYZHWJZDNCM-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

190.0 °F

flash_point_c

87.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service