Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

718076

Sigma-Aldrich

1-Bromo-3-chloro-2-propanol

96 %

Synonym(s):

1-Bromo-3-chloro-propan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6BrClO
CAS Number:
Molecular Weight:
173.44
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

liquid

refractive index

n20/D 1.519

capacity

96 %

density

1.785 g/mL at 25 °C

SMILES string

OC(CCl)CBr

InChI

1S/C3H6BrClO/c4-1-3(6)2-5/h3,6H,1-2H2

InChI key

TVDCSPSRGSLXOV-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorraine Richardson et al.
Reproduction (Cambridge, England) (2019-01-05)
The aim of this study was to investigate the properties and to functionally characterise the cervical mucus that modulates sperm transport through the cervix by using ewe breeds with a divergent pregnancy rate (Belclare and Suffolk; high and low, respectively)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service