Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

716642

Sigma-Aldrich

2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23OP
CAS Number:
Molecular Weight:
346.40
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22
form:
solid
assay:
97%

assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

106-110 °C

functional group

aldehyde
phosphine

SMILES string

Cc1cc(C)cc(c1)P(c2cc(C)cc(C)c2)c3ccccc3C=O

InChI

1S/C23H23OP/c1-16-9-17(2)12-21(11-16)25(22-13-18(3)10-19(4)14-22)23-8-6-5-7-20(23)15-24/h5-15H,1-4H3

InChI key

PQIAXRGPKDEYLW-UHFFFAOYSA-N

Application

Reactant for asymmetric hydrogenation

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service