Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

708429

Sigma-Aldrich

Ethyl 2-bromothiazole-4-carboxylate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrNO2S
CAS Number:
Molecular Weight:
236.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

powder

mp

67-71 °C

functional group

bromo
ester

storage temp.

2-8°C

SMILES string

CCOC(=O)c1csc(Br)n1

InChI

1S/C6H6BrNO2S/c1-2-10-5(9)4-3-11-6(7)8-4/h3H,2H2,1H3

InChI key

CNHISCQPKKGDPO-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service