Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

708372

Sigma-Aldrich

5-Amino-4-(4-bromophenyl)-3-methylpyrazole

96%

Synonym(s):

3-Amino-4-(4-bromophenyl)-2H-5-methylpyrazole, 4-(4-Bromophenyl)-5-methyl-2H-pyrazole-3-yl amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10BrN3
CAS Number:
Molecular Weight:
252.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

powder

mp

133-138 °C

functional group

bromo

SMILES string

Cc1n[nH]c(N)c1-c2ccc(Br)cc2

InChI

1S/C10H10BrN3/c1-6-9(10(12)14-13-6)7-2-4-8(11)5-3-7/h2-5H,1H3,(H3,12,13,14)

InChI key

XBGQAVPNQRLZOV-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service