Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

704075

Sigma-Aldrich

Bis(2,4,6-trimethylphenyl)phosphine

97%

Synonym(s):

Dimesitylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23P
CAS Number:
Molecular Weight:
270.35
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

95-100 °C

functional group

phosphine

SMILES string

Cc1cc(C)c(Pc2c(C)cc(C)cc2C)c(C)c1

InChI

1S/C18H23P/c1-11-7-13(3)17(14(4)8-11)19-18-15(5)9-12(2)10-16(18)6/h7-10,19H,1-6H3

InChI key

NBXSLXZWWHEDPI-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service