Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

702943

Sigma-Aldrich

Cy-vBRIDP

Synonym(s):

2-(Dicyclohexylphosphino)-1,1-diphenyl-1-propene, Dicyclohexyl(1-methyl-2,2-diphenylvinyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H35P
CAS Number:
Molecular Weight:
390.54
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

124-130 °C

functional group

phosphine

storage temp.

2-8°C

SMILES string

C\C(P(C1CCCCC1)C2CCCCC2)=C(\c3ccccc3)c4ccccc4

InChI

1S/C27H35P/c1-22(27(23-14-6-2-7-15-23)24-16-8-3-9-17-24)28(25-18-10-4-11-19-25)26-20-12-5-13-21-26/h2-3,6-9,14-17,25-26H,4-5,10-13,18-21H2,1H3

InChI key

YMSBPYCREGBACF-UHFFFAOYSA-N

Application

Catalyst for cycloaddition reactions

Legal Information

Sold in collaboration with Takasago for research purposes only. US6455720

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service