Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

702056

Sigma-Aldrich

3-Amino-4-(Boc-amino)pyridine

97%

Synonym(s):

(3-Aminopyridin-4-yl)-carbamic acid tert-butyl ester, 3-Amino-4-[(N-tert-butoxycarbonyl)amino]pyridine, 4-N-Boc-3,4-diaminopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N3O2
CAS Number:
Molecular Weight:
209.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

mp

123-127 °C

SMILES string

CC(C)(C)OC(=O)Nc1ccncc1N

InChI

1S/C10H15N3O2/c1-10(2,3)15-9(14)13-8-4-5-12-6-7(8)11/h4-6H,11H2,1-3H3,(H,12,13,14)

InChI key

UVAXTPFOVJAXHO-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service