Skip to Content
MilliporeSigma
All Photos(1)

Documents

700738

Sigma-Aldrich

N-Boc-7-azaindole

95%

Synonym(s):

1-(tert-Butoxycarbonyl)-1H-pyrrolo[2,3-b]pyridine, 1-(tert-Butoxycarbonyl)-7-azaindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.546

density

1.135 g/mL at 25 °C

SMILES string

CC(C)(C)OC(=O)n1ccc2cccnc12

InChI

1S/C12H14N2O2/c1-12(2,3)16-11(15)14-8-6-9-5-4-7-13-10(9)14/h4-8H,1-3H3

InChI key

SGZMGGUBXZBVPJ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service