Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

698288

Sigma-Aldrich

Di-tert-butylcyclohexylphosphine

95%

Synonym(s):

Cyclohexyldi-tert-butylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H29P
CAS Number:
Molecular Weight:
228.35
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

assay

95%

form

liquid

reaction suitability

reagent type: ligand

refractive index

n20/D 1.506

density

0.889 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)(C)P(C1CCCCC1)C(C)(C)C

InChI

1S/C14H29P/c1-13(2,3)15(14(4,5)6)12-10-8-7-9-11-12/h12H,7-11H2,1-6H3

InChI key

HFFHNJKBAYQARL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Dihydrogen activation
  • Synthesis of trinuclear ruthenium complexes

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

154.9 °F

flash_point_c

68.3 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service