Skip to Content
MilliporeSigma
All Photos(1)

Documents

697494

Sigma-Aldrich

4-Formylphenylboronic acid MIDA ester

96%

Synonym(s):

4-Formylphenylboronic acid methyliminoacetic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12BNO5
Molecular Weight:
261.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

powder

mp

242-248 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccc(C=O)cc2

InChI

1S/C12H12BNO5/c1-14-6-11(16)18-13(19-12(17)7-14)10-4-2-9(8-15)3-5-10/h2-5,8H,6-7H2,1H3

InChI key

ATAFRFKGTAHILE-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service