Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695866

Sigma-Aldrich

4-Methyl-3-pentenylboronic acid pinacol ester

97%

Synonym(s):

2-(4-Methyl-3-pentenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Methylpent-2-ene-5-boronic acid pinacol ester, 4,4,5,5-Tetramethyl-2-(4-methyl-3-pentenyl)-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23BO2
Molecular Weight:
210.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.441

bp

219-223 °C/760 mmHg

density

0.881 g/mL at 25 °C

SMILES string

C\C(C)=C\CCB1OC(C)(C)C(C)(C)O1

InChI

1S/C12H23BO2/c1-10(2)8-7-9-13-14-11(3,4)12(5,6)15-13/h8H,7,9H2,1-6H3

InChI key

WHSCXARPDWZZRD-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service