Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695513

Sigma-Aldrich

3-[(tert-Butyldimethylsilyloxy)methyl]aziridine-2-carboxaldehyde dimer

Synonym(s):

6-[(tert-Butyldimethylsilyloxy)methyl]-2-[3-[(tert-butyldimethylsilyloxy)methyl]-2-aziridinyl]-3-oxa-1-azabicyclo[3.1.0]hexan-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H42N2O4Si2
CAS Number:
Molecular Weight:
430.73
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.456

density

0.953 g/mL at 25 °C

SMILES string

CC(C)(C)[Si](C)(C)OCC1NC1C2OC(O)C3C(CO[Si](C)(C)C(C)(C)C)N23

InChI

1S/C20H42N2O4Si2/c1-19(2,3)27(7,8)24-11-13-15(21-13)17-22-14(16(22)18(23)26-17)12-25-28(9,10)20(4,5)6/h13-18,21,23H,11-12H2,1-10H3

InChI key

TVJBDLSJTDKBNQ-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

150.1 °F

flash_point_c

65.6 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service