Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

694401

Sigma-Aldrich

2,2-Dichloro-3,3,3-trifluoropropionic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$224.00

$224.00


Available to ship onApril 14, 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$224.00

About This Item

Empirical Formula (Hill Notation):
C3HCl2F3O2
CAS Number:
Molecular Weight:
196.94
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$224.00


Available to ship onApril 14, 2025Details


Request a Bulk Order

Quality Level

assay

97%

form

solid

bp

50-51 °C/15 mmHg

mp

26-34 °C

functional group

carboxylic acid
chloro
fluoro

SMILES string

OC(=O)C(Cl)(Cl)C(F)(F)F

InChI

1S/C3HCl2F3O2/c4-2(5,1(9)10)3(6,7)8/h(H,9,10)

Inchi Key

OEOQSGPFZFSHBS-UHFFFAOYSA-N

Pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dmitriy Ovcharenko et al.
Cancer biology & therapy, 20(9), 1281-1289 (2019-06-27)
Dichloroacetate (DCA) is an inhibitor of pyruvate dehydrogenase kinase (PDK) that has been shown to reverse the Warburg effect and cause tumor cell death. Clinical research into the anti-cancer activity of DCA revealed high dosage requirements and reports of toxicity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service