Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688711

Sigma-Aldrich

tert-Butyl crotonate

≥97.0%

Synonym(s):

tert-Butyl (E)-2-butenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0%

form

liquid

refractive index

n20/D 1.425

functional group

ester

storage temp.

2-8°C

SMILES string

C\C=C\C(=O)OC(C)(C)C

InChI

1S/C8H14O2/c1-5-6-7(9)10-8(2,3)4/h5-6H,1-4H3/b6-5+

InChI key

QHSPZGZEUDEIQM-AATRIKPKSA-N

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

73.4 °F

flash_point_c

23 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service