Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

686646

Sigma-Aldrich

Polyester-8-hydroxyl-1-acetylene bis-MPA dendron, generation 3

Synonym(s):

Dendron-G3-Acetylene-OH

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

hydroxyl surface groups

form

powder

mol wt

generation 3

feature

focal point acetylene

no. Surface Groups

8

storage temp.

2-8°C

SMILES string

CC(CO)(CO)C(=O)OCC(C)(COC(=O)C(C)(CO)CO)C(=O)OCC(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)C(=O)OCC#C

InChI

1S/C38H60O22/c1-9-10-54-29(51)36(6,19-59-30(52)37(7,21-55-25(47)32(2,11-39)12-40)22-56-26(48)33(3,13-41)14-42)20-60-31(53)38(8,23-57-27(49)34(4,15-43)16-44)24-58-28(50)35(5,17-45)18-46/h1,39-46H,10-24H2,2-8H3

InChI key

IBDJAKQCSBCQRU-UHFFFAOYSA-N

General description

calculated mol. wt. 868.9

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

212.0 °F - closed cup

flash_point_c

100.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dendrons in nanomaterials - Find features and benefits of polyester dendrons and hyperbranched polymer products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service