Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

685615

Sigma-Aldrich

Bromotricarbonyl(tetrahydrofuran)rhenium(I) dimer

Synonym(s):

Dibromohexacarbonylbis(tetrahydrofuran)dirhenium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[ReBr(CO)3(C4H8O)]2
CAS Number:
Molecular Weight:
844.49
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: rhenium
reagent type: catalyst

SMILES string

Br[Re].Br[Re].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].C1CCOC1.C2CCOC2

InChI

1S/2C4H8O.6CO.2BrH.2Re/c2*1-2-4-5-3-1;6*1-2;;;;/h2*1-4H2;;;;;;;2*1H;;/q;;;;;;;;;;2*+1/p-2

Inchi Key

GARJHXMUDDMPBU-UHFFFAOYSA-L

Application

Catalyst for insertation of acetylenes into 1,3-dicarbonyl′s and for isobenzofuran synthesis[1][2].

Pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoichiro Kuninobu et al.
Journal of the American Chemical Society, 128(38), 12376-12377 (2006-09-21)
A rhenium complex, [ReBr(CO)3(thf)]2, catalyzed reactions of aromatic ketimines with aldehydes to give isobenzofuran derivatives in good to excellent yields. In contrast to ruthenium and rhodium catalysts, aldehydes, which are polar unsaturated molecules, inserted into the C-H bond after activation
Takai, K. et al.
Journal of the American Chemical Society, 128, 11369-11369 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service