Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

683760

SAFC

2-[(Trimethylsilyl)ethynyl]benzonitrile

97%

Synonym(s):

1-(2-Cyanophenyl)-2-(trimethylsilyl)acetylene, 2-[(Trimethylsilanyl)ethynyl]benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NSi
CAS Number:
Molecular Weight:
199.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.537

density

0.942 g/mL at 25 °C

SMILES string

C[Si](C)(C)C#Cc1ccccc1C#N

InChI

1S/C12H13NSi/c1-14(2,3)9-8-11-6-4-5-7-12(11)10-13/h4-7H,1-3H3

InChI key

OSJJQDZTHSMKBV-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service