Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

681253

Sigma-Aldrich

Sodium nitromalonaldehyde monohydrate

97%

Synonym(s):

(1-Formyl-1-nitro-2-oxoethyl) sodium monohydrate, 3-Hydroxy-2-nitro-2-propenal sodium salt monohydrate, 3-Hydroxy-2-nitroacrolein sodium salt monohydrate, Sodium nitromalondialdehyde monohydrate, Sodium nitromalonic dialdehyde monohydrate, Sodium nitropropanedial monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2NNaO4 · H2O
CAS Number:
Molecular Weight:
157.06
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

120-124 °C

functional group

aldehyde
amine
nitro

storage temp.

2-8°C

SMILES string

O.[Na+].[O-][N+](=O)[C-](C=O)C=O

InChI

1S/C3H2NO4.Na.H2O/c5-1-3(2-6)4(7)8;;/h1-2H;;1H2/q-1;+1;

InChI key

DQQCQHYLSWEYSN-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service