Recommended Products
assay
97%
refractive index
n20/D 1.5618
density
1.4869 g/mL at 25 °C
SMILES string
Cc1cncc(Br)c1
InChI
1S/C6H6BrN/c1-5-2-6(7)4-8-3-5/h2-4H,1H3
InChI key
ADCLTLQMVAEBLB-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
195.1 °F - closed cup
flash_point_c
90.6 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service