Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67948

Sigma-Aldrich

(R)-Tetrahydrofuran-2-carbonitrile

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(R)-Tetrahydrofurfurylnitrile, (R)-Tetrahydrofuroic nitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO
CAS Number:
Molecular Weight:
97.12
Beilstein/REAXYS Number:
7197285
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (sum of enantiomers, GC)

optical purity

enantiomeric ratio: ≥99:1 (GC)

SMILES string

N#C[C@H]1CCCO1

InChI

1S/C5H7NO/c6-4-5-2-1-3-7-5/h5H,1-3H2/t5-/m1/s1

InChI key

RZZIKHSWRWWPAN-RXMQYKEDSA-N

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

1.4 °F

flash_point_c

-17 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service