675016
1-(2-Hydroxyphenyl)piperazine
97%
Synonym(s):
2-(1-Piperazino)phenol
About This Item
Recommended Products
assay
97%
mp
125-129 °C
SMILES string
Oc1ccccc1N2CCNCC2
InChI
1S/C10H14N2O/c13-10-4-2-1-3-9(10)12-7-5-11-6-8-12/h1-4,11,13H,5-8H2
InChI key
UORNTHBBLYBAJJ-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service