Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

674729

Sigma-Aldrich

3-(tert-Butyldimethylsilyloxy)-1-butyn-1-ylboronic acid pinacol ester

96%

Synonym(s):

2-((3-tert-Butyldimethylsilyloxy)-1-butyn-1-yl)-4,4,5,5-tetramethyl-(1,3,2)dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H31BO3Si
Molecular Weight:
310.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.4488

density

0.9158 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(O[Si](C)(C)C(C)(C)C)C#CB1OC(C)(C)C(C)(C)O1

InChI

1S/C16H31BO3Si/c1-13(18-21(9,10)14(2,3)4)11-12-17-19-15(5,6)16(7,8)20-17/h13H,1-10H3

InChI key

KGDJKWYVGUGRCG-UHFFFAOYSA-N

Application

Alkynylboronates participate in a variety of regio- and stereoselective carbon-carbon bond forming
processes including enyne cross-metathesis and the Alder ene reaction.[1][2]

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mansuk Kim et al.
Organic letters, 7(9), 1865-1868 (2005-04-23)
[reaction: see text] Regio- and stereoselective enyne cross metathesis reactions between borylated alkynes and terminal alkenes were realized to provide a variety of functionalized vinyl boronates. High chemical yield and regioselectivity was achieved irrespective of substituents on the alkyne and
Eric C Hansen et al.
Journal of the American Chemical Society, 127(10), 3252-3253 (2005-03-10)
Ruthenium-catalyzed Alder ene reactions between borylated alkynes and terminal alkenes give the corresponding beta,beta-disubstituted vinyl boronates with high selectivity for the branched isomer. The stereochemistry of the vinyl boronate moiety was the result of a formal trans addition of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service