672912
S-Butyl chlorothioformate
96%
Synonym(s):
(Butylthio)carbonyl chloride
About This Item
assay
96%
refractive index
n20/D 1.479
bp
49 °C/0.7 mmHg
density
0.985 g/mL
SMILES string
CCCCSC(Cl)=O
InChI
1S/C5H9ClOS/c1-2-3-4-8-5(6)7/h2-4H2,1H3
Inchi Key
YGLZPZAKCIKQKR-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service