Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

668532

Sigma-Aldrich

Di-μ-chlorobis[5-hydroxy-2-[1-(hydroxyimino-κN)ethyl]phenyl-κC]palladium(II) dimer

98%

Synonym(s):

Nájera Catalyst II, Di-μ-chloro-bis-[5-hydroxy-2-[1-(hydroxyimino-κN)-ethyl]-phenyl-κC]-palladium(II) dimer, Di-μ-chlorobis[5-hydroxy-2-[1-(hydroxyimino-κN)ethyl]phenyl-κC]dipalladium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd2(C8H8ClNO2)2
CAS Number:
Molecular Weight:
584.06
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

251-255 °C

SMILES string

C\C(=N/O)c1ccc(O)cc1[Pd]Cl.C\C(=N/O)c2ccc(O)cc2[Pd]Cl

InChI

1S/2C8H8NO2.2ClH.2Pd/c2*1-6(9-11)7-2-4-8(10)5-3-7;;;;/h2*2,4-5,10-11H,1H3;2*1H;;/q;;;;2*+1/p-2/b2*9-6+;;;;

InChI key

VEJSMXSVBZMTSO-FIOBSCOQSA-L

Application

Promotes the vinylation of aryl bromides and chlorides with vinylsiloxanes in the presence of aqueous NaOH.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Synthesis & Catalysis, 348, 2085-2085 (2006)

Articles

Palladacyclic catalysts by Bedford's group facilitate coupling reactions with low catalyst loadings and challenging aryl chlorides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service