Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

665967

Sigma-Aldrich

6-Chloropyridine-2-carbonitrile

96%

Synonym(s):

2-Chloro-6-cyanopyridine, 6-Chloropicolinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3ClN2
CAS Number:
Molecular Weight:
138.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

form

solid

mp

85-89 °C

SMILES string

Clc1cccc(n1)C#N

InChI

1S/C6H3ClN2/c7-6-3-1-2-5(4-8)9-6/h1-3H

InChI key

PGZHSVWXFKKCNR-UHFFFAOYSA-N

Application

Substrate used in a Negishi coupling reaction with an arylzinc halide catalyzed by Pd-NHC (PEPPSI-Ipr, Catalog No. 668032).[1]

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael G Organ et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(18), 4749-4755 (2006-03-29)
We have developed the first user-friendly Negishi protocol capable of routinely cross-coupling all combinations of alkyl and aryl centers. The use of an easily synthesized, air stable, highly active, well-defined precatalyst PEPPSI-IPr (1; PEPPSI=pyridine-enhanced precatalyst preparation, stabilization and initiation; IPr=diisopropylphenylimidazolium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service