Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

665770

Sigma-Aldrich

2,5-Bis(chloromethyl)-1,4-dimethoxybenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H12Cl2O2
CAS Number:
Molecular Weight:
235.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

mp

161-165 °C

SMILES string

COc1cc(CCl)c(OC)cc1CCl

Application

Anodic oxidation at a platinum electrode provides the corresponding bis(chloromethyl)quinone.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-Chu Zeng et al.
The Journal of organic chemistry, 69(4), 1053-1059 (2004-02-14)
The electrochemical oxidation of mono- and disubstituted 1,4-dimethoxybenzene derivatives with zero, one, and two benzylic CH(2)X groups (X = OAc, Cl, OH) (5a-c and 6a-c) has been carried out by using both constant-current and controlled-potential techniques in methanol and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service