Skip to Content
MilliporeSigma
All Photos(1)

Documents

665428

Sigma-Aldrich

Oxalyl-13C2 chloride

99 atom % 13C, 98% (CP)

Synonym(s):

Ethanedioyl-1,2-13C2 dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2Cl2O2
CAS Number:
Molecular Weight:
128.91
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Quality Level

assay

98% (CP)

bp

62-65 °C

density

1.523 g/mL at 25 °C

mass shift

M+2

storage temp.

2-8°C

SMILES string

Cl[13C](=O)[13C](Cl)=O

InChI

1S/C2Cl2O2/c3-1(5)2(4)6/i1+1,2+1

InChI key

CTSLXHKWHWQRSH-ZDOIIHCHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service