Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

655864

Sigma-Aldrich

5-(Aminomethyl)indole

95%

Synonym(s):

Indole-5-methanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2
CAS Number:
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

114-122 °C

functional group

amine

SMILES string

NCc1ccc2[nH]ccc2c1

InChI

1S/C9H10N2/c10-6-7-1-2-9-8(5-7)3-4-11-9/h1-5,11H,6,10H2

InChI key

UAYYSAPJTRVEQA-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • HIV-1 integrase inhibitors targeting the catalytic domain and its ability for interaction with LEDGF/p75
  • Inhibitors of Gli1-mediated transcription in the Hedgehog pathway
  • SCIO-469-like compounds for the inhibition of p38 MAP kinase
  • 4-(indol-5-ylamino)thieno[2,3-b]pyridine-5-carbonitriles as protein kinase c theta (PKC θ) inhibitors
  • Non-covalent thrombin inhibitors

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Farcaş et al.
Analytica chimica acta, 1034, 214-222 (2018-09-09)
Only few reports describe the use of capillary electrophoresis in the context of Fragment Based Drug Discovery (FBDD). In this paper, we will present a generic, fully automated, microscale electrophoretic mobility shift displacement assay that can be used in FBDD

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service