Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

654973

Sigma-Aldrich

Perfluorotridecanoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C(CF2)11CO2H
CAS Number:
Molecular Weight:
664.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

112-123 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C13HF25O2/c14-2(15,1(39)40)3(16,17)4(18,19)5(20,21)6(22,23)7(24,25)8(26,27)9(28,29)10(30,31)11(32,33)12(34,35)13(36,37)38/h(H,39,40)

InChI key

LVDGGZAZAYHXEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

target_organs

Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Cao et al.
The Science of the total environment, 751, 142255-142255 (2020-11-14)
Confident elemental composition determination of compounds in complex samples such as natural organic matter (NOM) by ultrahigh resolution Fourier transform ion cyclotron resonance mass spectrometry (FTICR-MS) is challenging due to the interference between multiple components in these samples during detection.
Jihyun Kim et al.
Chemosphere, 262, 128012-128012 (2020-11-14)
Due to global restriction on perfluorooctanesulfonic acid (PFOS) and perfluorooctanoic acid (PFOA), the use of long-chain perfluoroalkyl substances (PFASs, C > 8) and their environmental occurrences have increased. PFOS and PFOA have been known for thyroid disruption, however, knowledge is still limited
Yukiko Fujii et al.
Toxics, 8(2) (2020-04-05)
Perfluoroalkyl carboxylic acids (PFCAs), such as perfluorooctanoic acid (PFOA, C8), are a group of industrial chemicals that are detected in the serum of people throughout the world. Long-chain PFCAs (C9 to C13) have high lipophilicity, therefore they may have a
Sibel Barisci et al.
Chemosphere, 243, 125349-125349 (2019-11-23)
This study investigates electrooxidation of short (C3-C6) and long (C7-C-18) chain perfluorocarboxylic acids (PFCAs) including perfluorooctane sulfonate (PFOA) using Si/BDD electrode. The effect of operational parameters (supporting electrolyte type, applied current density, and initial pH) were explored for PFOA removal.
Karina Dale et al.
Aquatic toxicology (Amsterdam, Netherlands), 227, 105590-105590 (2020-09-06)
The aim of the present study was to investigate effects of defined mixtures of polycyclic aromatic hydrocarbons (PAHs) and perfluoroalkyl substances (PFASs), at low, environmentally relevant (1× = L), or high (20× = H) doses, on biological responses in Atlantic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service