Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648752

Sigma-Aldrich

4-Bromo-2-chloro-6-methylaniline

98%

Synonym(s):

4-Bromo-6-chloro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7BrClN
CAS Number:
Molecular Weight:
220.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

38-42 °C (lit.)

functional group

bromo
chloro

SMILES string

Cc1cc(Br)cc(Cl)c1N

InChI

1S/C7H7BrClN/c1-4-2-5(8)3-6(9)7(4)10/h2-3H,10H2,1H3

InChI key

DIXGIKZIIZRFKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service