Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

646806

Sigma-Aldrich

3-Chloro-6-fluorobenzo[b]thiophene-2-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4ClFO2S
CAS Number:
Molecular Weight:
230.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

293-297 °C (lit.)

functional group

carboxylic acid
chloro
fluoro

SMILES string

OC(=O)c1sc2cc(F)ccc2c1Cl

InChI

1S/C9H4ClFO2S/c10-7-5-2-1-4(11)3-6(5)14-8(7)9(12)13/h1-3H,(H,12,13)

Inchi Key

HSRSWUJIPYUCSE-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service